2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Acetoxyethyl)-4-methylthiazole, 4-Methyl-5-thiazolylethyl acetate

M1848-5G M1848-25G
TCM1848-5GEA 26.86 CAD
TCM1848-5G TCM1848-25G
2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98.0%
2-(4-Methyl-5-thiazolyl)ethyl acetate

CAS Number: 656-53-1
MDL Number: MFCD00005338
Molecular Formula: C8H11NO2S
Molecular Weight: 185.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 113
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: Acetic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Acetoxyethyl)-4-methylthiazole

Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
Boiling Pt: 118 °C
Density: 1.17 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00005338
CAS Number: 656-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=504fe840d799e159d4b72ed166e8b1510890f381&vid=504fe840d799e159d4b72ed166e8b1510890f381&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR