2-Methyl-3-tetrahydrothiophenone ≥97.0%

Supplier: TCI America

M1839-25G M1839-5G
TCM1839-25GEA 399.74 CAD
TCM1839-25G TCM1839-5G
2-Methyl-3-tetrahydrothiophenone ≥97.0%
2-Methyl-3-tetrahydrothiophenone

CAS Number: 13679-85-1
MDL Number: MFCD00078280
Molecular Formula: C5H8OS
Molecular Weight: 116.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 71
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 2-Methyl-3-oxotetrahydrothiophene

Formula: C₅H₈OS
Boiling Pt: 86 °C
Density: 1.12 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00078280
CAS Number: 13679-85-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=178c071c6acf47ab9c877c3090a7579e362c1ef3&vid=178c071c6acf47ab9c877c3090a7579e362c1ef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR