2-(Methylthio)pyrazine ≥98.0%

Supplier: TCI America

M1821-25G M1821-5G
TCM1821-25GEA 75.84 CAD
TCM1821-25G TCM1821-5G
2-(Methylthio)pyrazine ≥98.0%
2-(Methylthio)pyrazine

CAS Number: 21948-70-9
MDL Number: MFCD00041024
Molecular Formula: C5H6N2S
Molecular Weight: 126.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45

Formula: C₅H₆N₂S
Melting Pt: 45 °C
MDL Number: MFCD00041024
CAS Number: 21948-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd5153c37814f3a323c60c008ed99e97f637d5a2&vid=fd5153c37814f3a323c60c008ed99e97f637d5a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR