Methyl-4-methoxysalicylate ≥99.0%
Supplier: TCI America
M1797-25G
M1797-5G
TCM1797-25GEA
143.78
CAD
TCM1797-25G
TCM1797-5G
Methyl-4-methoxysalicylate ≥99.0%
Methyl 4-methoxysalicylate
CAS Number: 5446-02-6
MDL Number: MFCD00008424
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 50
Flash Point (°C): 113
Other Product Names / Synonyms: 4-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-4-methoxybenzoate, 2-Hydroxy-4-methoxybenzoic Acid Methyl Ester, Methyl 2-Hydroxy-p-anisate
Formula:
C₉H₁₀O₄ Boiling Pt: 134 °C Melting Pt: 50 °C Flash Pt: 113 °C |
MDL Number:
MFCD00008424 CAS Number: 5446-02-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...