Methyl-2,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1796-25G
M1796-5G
TCM1796-25GEA
118.5
CAD
TCM1796-25G
TCM1796-5G
Methyl-2,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl 2,4-dihydroxybenzoate
CAS Number: 2150-47-2
MDL Number: MFCD00002276
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 119
Other Product Names / Synonyms: 2,4-Dihydroxybenzoic Acid Methyl Ester, Methyl beta-Resorcylate, beta-Resorcylic Acid Methyl Ester
Formula:
C₈H₈O₄ Melting Pt: 119 °C |
MDL Number:
MFCD00002276 CAS Number: 2150-47-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...