trans-4-Methylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1783-25G M1783-5G
TCM1783-25GEA 355.34 CAD
TCM1783-25G TCM1783-5G
trans-4-Methylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Methylcyclohexanecarboxylic acid

CAS Number: 13064-83-0
MDL Number: MFCD00074943
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 245
Melting point (°C): 112

Formula: C₈H₁₄O₂
Boiling Pt: 245 °C
Melting Pt: 112 °C
MDL Number: MFCD00074943
CAS Number: 13064-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e52b014119531c9bbdd967c689c8de487ac7c227&vid=e52b014119531c9bbdd967c689c8de487ac7c227&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR