Methyl-4-chloro-3-nitrobenzoate ≥98.0%

Supplier: TCI America

M1779-25G M1779-5G
TCM1779-25GEA 157.05 CAD
TCM1779-25G TCM1779-5G
Methyl-4-chloro-3-nitrobenzoate ≥98.0%
Methyl 4-chloro-3-nitrobenzoate

CAS Number: 14719-83-6
MDL Number: MFCD00016989
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Other Product Names / Synonyms: 4-Chloro-3-nitrobenzoic Acid Methyl Ester

Formula: C₈H₆ClNO₄
Melting Pt: 82 °C
MDL Number: MFCD00016989
CAS Number: 14719-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec0db164a4d52e9e5c0cfd66ebfa10f05e0c60c&vid=8ec0db164a4d52e9e5c0cfd66ebfa10f05e0c60c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR