Methyl indole-4-carboxylate ≥98.0%

Supplier: TCI America

M1773-5G M1773-1G
TCM1773-5GEA 180.12 CAD
TCM1773-5G TCM1773-1G
Methyl indole-4-carboxylate ≥98.0%
Methyl indole-4-carboxylate

CAS Number: 39830-66-5
MDL Number: MFCD00191222
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 71


Other Product Names / Synonyms: Indole-4-carboxylic Acid Methyl Ester

Formula: C₁₀H₉NO₂
MW: 175.19
Melting Pt: 71 °C
MDL Number: MFCD00191222
CAS Number: 39830-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=983bf379ca56fd9f321d55b53cc94f0dfe6320ae&vid=983bf379ca56fd9f321d55b53cc94f0dfe6320ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR