Methyl-3-amino-4-methylthiophene-2-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 2-Thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, Methyl 3-amino-4-methylthiophene-2-carboxylate

M1772-5G M1772-25G
TCM1772-5GEA 47.4 CAD
TCM1772-5G TCM1772-25G
Methyl-3-amino-4-methylthiophene-2-carboxylate ≥98.0%
Methyl-3-amino-4-methylthiophene-2-carboxylate

CAS Number: 85006-31-1
MDL Number: MFCD00051822
Molecular Formula: C7H9NO2S
Molecular Weight: 171.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86
Flash Point (°C): 100


Other Product Names / Synonyms: 3-Amino-4-methylthiophene-2-carboxylic Acid Methyl Ester

Formula: C₇H₉NO₂S
MW: 171.21 g/mol
Melting Pt: 86 °C
Flash Pt: 100 °C
MDL Number: MFCD00051822
CAS Number: 85006-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b4e05fe7475af43845563d24b09fa3bf2b43d3c&vid=6b4e05fe7475af43845563d24b09fa3bf2b43d3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR