Methyl vanillate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 4-hydroxy-3-methoxybenzoate
M1738-25G
M1738-5G
TCM1738-25GEA
159.58
CAD
TCM1738-25G
TCM1738-5G
Methyl vanillate ≥98.0%
Methyl vanillate
CAS Number: 3943-74-6
MDL Number: MFCD00008438
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 287
Melting point (°C): 65
Other Product Names / Synonyms: Vanillic Acid Methyl Ester, Methyl 4-Hydroxy-3-methoxybenzoate, 4-Hydroxy-3-methoxybenzoic Acid Methyl Ester
Formula:
C₉H₁₀O₄ Boiling Pt: 287 °C Melting Pt: 65 °C |
MDL Number:
MFCD00008438 CAS Number: 3943-74-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...