Methyl-2-chloro-4-nitrobenzoate ≥99.0%

Supplier: TCI America

Synonyms: 2-Chloro-4-nitrobenzoic acid methyl ester, Methyl 2-chloro-4-nitrobenzoate

M1724-25G M1724-5G
TCM1724-25GEA 251.22 CAD
TCM1724-25G TCM1724-5G
Methyl-2-chloro-4-nitrobenzoate ≥99.0%
Methyl 2-chloro-4-nitrobenzoate

CAS Number: 13324-11-3
MDL Number: MFCD00017013
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 77


Other Product Names / Synonyms: 2-Chloro-4-nitrobenzoic Acid Methyl Ester

Formula: C₈H₆ClNO₄
Melting Pt: 77 °C
MDL Number: MFCD00017013
CAS Number: 13324-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f5562c911f1f5d805cdf533cc7c93cc143d5dd&vid=e2f5562c911f1f5d805cdf533cc7c93cc143d5dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR