7-Methoxycoumarin ≥98.0%

Supplier: TCI America

M1723-25G M1723-5G
TCM1723-25GEA 698.36 CAD
TCM1723-25G TCM1723-5G
7-Methoxycoumarin ≥98.0%
7-Methoxycoumarin

CAS Number: 531-59-9
MDL Number: MFCD00006876
Molecular Formula: C10H8O3
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystalline Powder
Melting point (°C): 118

Formula: C₁₀H₈O₃
Melting Pt: 118 °C
MDL Number: MFCD00006876
CAS Number: 531-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34066db862e23e8ad3e486e143e914f8fb52bf74&vid=34066db862e23e8ad3e486e143e914f8fb52bf74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR