5-Nitro-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1716-1G M1716-5G
TCM1716-1GEA 28.44 CAD
TCM1716-1G TCM1716-5G
5-Nitro-o-toluic acid ≥98.0% (by GC, titration analysis)
5-Nitro-o-toluic acid

CAS Number: 1975-52-6
MDL Number: MFCD00007371
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 178


Other Product Names / Synonyms: 5-Nitro-o-toluic Acid

Formula: C₈H₇NO₄
Melting Pt: 178 °C
MDL Number: MFCD00007371
CAS Number: 1975-52-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15d104c4f3ed1899ec91625f8219880cfa1a1357&vid=94ba9a73790e26aff63ea8fd17ba3195a23e20b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR