2-(Methylthio)thiazole ≥98.0%

Supplier: TCI America

M1712-5G M1712-25G
TCM1712-5GEA 77.42 CAD
TCM1712-5G TCM1712-25G
2-(Methylthio)thiazole ≥98.0%
2-(Methylthio)thiazole

CAS Number: 5053-24-7
Molecular Formula: C4H5NS2
Molecular Weight: 131.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Flash Point (°C): 91
Specific Gravity (20/20): 1.28

Formula: C₄H₅NS₂
Boiling Pt: 62 °C
Density: 1.28 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00858981
CAS Number: 5053-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eb4d247e3e3f50e67a79c77f4ceb28b2f0bfbbc&vid=2eb4d247e3e3f50e67a79c77f4ceb28b2f0bfbbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR