5-Methylisatin ≥99.0%

Supplier: TCI America

Synonyms: 5-Methylindoline-2,3-dione, 5-Methyl-1H-indole-2,3-dione

M1703-25G M1703-5G
TCM1703-25GEA 202.24 CAD
TCM1703-25G TCM1703-5G
5-Methylisatin ≥99.0%
5-Methylisatin

CAS Number: 608-05-9
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 188


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 5-Methylindole-2,3-dione, 5-Methyl-2,3-indolinedione

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00005721
CAS Number: 608-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60c705a27d96528b65bd6159cf6bcf3eb0e7144b&vid=60c705a27d96528b65bd6159cf6bcf3eb0e7144b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR