7-Methylindole ≥98.0%

Supplier: TCI America

M1702-1G M1702-5G
TCM1702-1GEA 85.32 CAD
TCM1702-1G TCM1702-5G
7-Methylindole ≥98.0%
7-Methylindole

CAS Number: 933-67-5
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 266
Melting point (°C): 83

Formula: C₉H₉N
Boiling Pt: 266 °C
Melting Pt: 83 °C
MDL Number: MFCD00005684
CAS Number: 933-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a2d75d18bb6d4a344dadda58080951869a89be2&vid=8a2d75d18bb6d4a344dadda58080951869a89be2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR