4-Methoxypyridine ≥98.0%

Supplier: TCI America

M1696-25G M1696-5G
TCM1696-25GEA 196.05 CAD
TCM1696-25G TCM1696-5G
4-Methoxypyridine ≥98.0%
4-Methoxypyridine

CAS Number: 620-08-6
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 190
Melting point (°C): 4
Flash Point (°C): 74
Specific Gravity (20/20): 1.08

Formula: C₆H₇NO
Boiling Pt: 190 °C
Melting Pt: 4 °C
Density: 1.08 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00674049
CAS Number: 620-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bea1a64bf22e647f7a2b6ece5ea7a0a072f8262&vid=8bea1a64bf22e647f7a2b6ece5ea7a0a072f8262&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR