Methyl-4-tert-butylbenzoate ≥98.0%

Supplier: TCI America

M1693-500G M1693-25G
TCM1693-500GEA 180.12 CAD
TCM1693-500G TCM1693-25G
Methyl-4-tert-butylbenzoate ≥98.0%
Methyl 4-tert-butylbenzoate

CAS Number: 26537-19-9
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 4-tert-Butylbenzoic Acid Methyl Ester

Formula: C₁₂H₁₆O₂
Boiling Pt: 247 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00008835
CAS Number: 26537-19-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20e45d3e3028c063468cc183dc4d5b0d6f0cd140&vid=20e45d3e3028c063468cc183dc4d5b0d6f0cd140&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR