5-Methylbenzoxazole ≥98.0%

Supplier: TCI America

M1688-5G M1688-25G
TCM1688-5GEA 262.28 CAD
TCM1688-5G TCM1688-25G
5-Methylbenzoxazole ≥98.0%
5-Methylbenzoxazole

CAS Number: 10531-78-9
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 45

Formula: C₈H₇NO
Boiling Pt: 96 °C
Melting Pt: 45 °C
MDL Number: MFCD00216938
CAS Number: 10531-78-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3275520e07c8d895cc9e5fa7131c048970439af6&vid=3275520e07c8d895cc9e5fa7131c048970439af6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR