4'-(Methylthio)acetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-(Methylthio)phenyl)ethanone

M1672-25G M1672-5G
TCM1672-25GEA 307.47 CAD
TCM1672-25G TCM1672-5G
4'-(Methylthio)acetophenone ≥98.0%
4'-(Methylthio)acetophenone

CAS Number: 1778-09-2
Molecular Formula: C9H10OS
Molecular Weight: 166.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 79


Other Product Names / Synonyms: 4-(Acetyl)thioanisole

Formula: C₉H₁₀OS
MW: 166.24 g/mol
Boiling Pt: 135 °C
Melting Pt: 79 °C
MDL Number: MFCD00039835
CAS Number: 1778-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31b5ae8b2b0a243a3074ad9589e3fdb37ca5dd72&vid=31b5ae8b2b0a243a3074ad9589e3fdb37ca5dd72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR