2-Methoxythiophenol ≥98.0%
Supplier: TCI America
M1670-25G
M1670-5G
TCM1670-25GEA
440.95
CAD
TCM1670-25G
TCM1670-5G
2-Methoxythiophenol ≥98.0%
2-Methoxythiophenol
CAS Number: 7217-59-6
Molecular Formula: C7H8OS
Molecular Weight: 140.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Flash Point (°C): 95
Specific Gravity (20/20): 1.15
Other Product Names / Synonyms: 2-Mercaptoanisole, 2-Methoxythiophenol
Formula:
C₇H₈OS Boiling Pt: 219 °C Density: 1.15 (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00004834 CAS Number: 7217-59-6 UN: 3334 ADR: 9, |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...