Methyl-5-acetylsalicylate ≥98.0%

Supplier: TCI America

M1658-25G M1658-500G
TCM1658-25GEA 55.3 CAD
TCM1658-25G TCM1658-500G
Methyl-5-acetylsalicylate ≥98.0%
Methyl 5-acetylsalicylate

CAS Number: 16475-90-4
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 167
Melting point (°C): 60


Other Product Names / Synonyms: 5-Acetyl-2-hydroxybenzoic Acid Methyl Ester, 5-Acetylsalicylic Acid Methyl Ester, Methyl 5-Acetyl-2-hydroxybenzoate

Formula: C₁₀H₁₀O₄
Boiling Pt: 167 °C
Melting Pt: 60 °C
MDL Number: MFCD00040924
CAS Number: 16475-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd581240829d8f85dfbede0e51797e13193e2df8&vid=dd581240829d8f85dfbede0e51797e13193e2df8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR