2-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M1570-1G
M1570-5G
TCM1570-1GEA
74.26
CAD
TCM1570-1G
TCM1570-5G
2-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
2-(Methylthio)phenylboronic acid
CAS Number: 168618-42-6
MDL Number: MFCD01318165
Molecular Formula: C7H9BO2S
Molecular Weight: 168.02
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 83
Other Product Names / Synonyms: 2-(Methylthio)benzeneboronic Acid
Formula:
C₇H₉BO₂S Melting Pt: 83 °C |
MDL Number:
MFCD01318165 CAS Number: 168618-42-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...