Methylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M1553-1G
M1553-5G
TCM1553-1GEA
63.2
CAD
TCM1553-1G
TCM1553-5G
Methylboronic acid (contains varying amounts of Anhydride)
Methylboronic acid
CAS Number: 13061-96-6
MDL Number: MFCD00002105
Molecular Formula: CH5BO2
Molecular Weight: 59.86
Form: Crystal
Color: White
Melting point (°C): 91
Other Product Names / Synonyms: Methaneboronic Acid
Formula:
CH₅BO₂ Melting Pt: 91 °C |
MDL Number:
MFCD00002105 CAS Number: 13061-96-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...