6-Methoxy-1-indanone ≥98.0%

Supplier: TCI America

M1535-5G M1535-25G
TCM1535-5GEA 56.88 CAD
TCM1535-5G TCM1535-25G
6-Methoxy-1-indanone ≥98.0%
6-Methoxy-1-indanone

CAS Number: 13623-25-1
MDL Number: MFCD00021232
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 109

Formula: C₁₀H₁₀O₂
Melting Pt: 109 °C
MDL Number: MFCD00021232
CAS Number: 13623-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9170437cfa14c2999f5b410f452d188f1282771&vid=a9170437cfa14c2999f5b410f452d188f1282771&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR