Methyl-4-(trifluoromethyl)benzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-(trifluoromethyl)benzoate

M1498-25G M1498-5G
TCM1498-25GEA 156.42 CAD
TCM1498-25G TCM1498-5G
Methyl-4-(trifluoromethyl)benzoate ≥98.0%
Methyl 4-(trifluoromethyl)benzoate

CAS Number: 2967-66-0
MDL Number: MFCD00042324
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 82
Specific Gravity (20/20): 1.30


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Methyl alpha,alpha,alpha-Trifluoro-p-toluate, 4-(Trifluoromethyl)benzoic Acid Methyl Ester

Formula: C₉H₇F₃O₂
Density: 1.30 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00042324
CAS Number: 2967-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfcde5abf21f927b99375e3c388b7dd66ded09fb&vid=dfcde5abf21f927b99375e3c388b7dd66ded09fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR