5-Methoxy-1-indanone ≥98.0%

Supplier: TCI America

M1467-25G M1467-5G
TCM1467-25GEA 323.9 CAD
TCM1467-25G TCM1467-5G
5-Methoxy-1-indanone ≥98.0%
5-Methoxy-1-indanone

CAS Number: 5111-70-6
MDL Number: MFCD00003789
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 112

Formula: C₁₀H₁₀O₂
Boiling Pt: 150 °C
Melting Pt: 112 °C
MDL Number: MFCD00003789
CAS Number: 5111-70-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2126f01f236b4899124472422f85a778b0b5e4&vid=ec2126f01f236b4899124472422f85a778b0b5e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR