3-Methylphthalic anhydride ≥96.0%

Supplier: TCI America

Synonyms: 4-Methylisobenzofuran-1,3-dione

M1464-1G M1464-5G
TCM1464-1GEA 72.68 CAD
TCM1464-1G TCM1464-5G
3-Methylphthalic anhydride ≥96.0%
3-Methylphthalic anhydride

CAS Number: 4792-30-7
Molecular Formula: C9H6O3
Molecular Weight: 162.14
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 118

Formula: C₉H₆O₃
MW: 162.14
Melting Pt: 118 °C
MDL Number: MFCD00047316
CAS Number: 4792-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b81fa1c5f9c4a6896add8ed06694f03c618007e9&vid=b81fa1c5f9c4a6896add8ed06694f03c618007e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR