3-Methyl-2-thenoic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Methylthiophene-2-carboxylic acid

M1459-25G M1459-5G
TCM1459-25GEA 80.58 CAD
TCM1459-25G TCM1459-5G
3-Methyl-2-thenoic acid ≥98.0%
3-Methyl-2-thenoic acid

CAS Number: 23806-24-8
MDL Number: MFCD00005438
Molecular Formula: C6H6O2S
Molecular Weight: 142.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 3-Methyl-2-thenoic Acid

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00005438
CAS Number: 23806-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d955ed8a54ab73068142d01494643c0a28ce071&vid=9d955ed8a54ab73068142d01494643c0a28ce071&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR