4-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M1458-1G
M1458-5G
TCM1458-1GEA
42.66
CAD
TCM1458-1G
TCM1458-5G
4-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
4-(Methylthio)phenylboronic acid
CAS Number: 98546-51-1
MDL Number: MFCD00093410
Molecular Formula: C7H9BO2S
Molecular Weight: 168.02
Form: Crystal
Color: White
Melting point (°C): 213
Other Product Names / Synonyms: 4-(Methylthio)benzeneboronic Acid
Formula:
C₇H₉BO₂S Melting Pt: 213 °C |
MDL Number:
MFCD00093410 CAS Number: 98546-51-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...