3-Methyl-2-pyridone ≥98.0%
Supplier: TCI America
Synonyms:
3-Methyl-2-pyridinone, 2-Hydroxy-3-methylpyridine, 3-Methylpyridin-2(1H)-one
M1447-5G
M1447-25G
TCM1447-5GEA
197.5
CAD
TCM1447-5G
TCM1447-25G
3-Methyl-2-pyridone ≥98.0%
3-Methyl-2-pyridone
CAS Number: 1003-56-1
MDL Number: MFCD00039704
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Other Product Names / Synonyms: 2-Hydroxy-3-methylpyridine, 3-Methyl-2-pyridinone
Formula:
C₆H₇NO MW: 109.13 |
MDL Number:
MFCD00039704 CAS Number: 1003-56-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...