1-Methyl-3-propylimidazolium iodide ≥97.0%
Supplier: TCI America
Synonyms:
1-Methyl-3-n-propylimidazolium iodide, 1-Methyl-3-propyl-1H-imidazol-3-ium iodide
M1440-25G
M1440-5G
TCM1440-25GEA
150.1
CAD
TCM1440-25G
TCM1440-5G
1-Methyl-3-propylimidazolium iodide ≥97.0%
1-Methyl-3-propylimidazolium iodide
CAS Number: 119171-18-5
Molecular Formula: C7H13IN2
Molecular Weight: 252.10
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Clear Liquid
Formula:
C₇H₁₃IN₂ |
MDL Number:
MFCD08276385 CAS Number: 119171-18-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...