2'-Methylformanilide ≥98.0%

Supplier: TCI America

M1431-25G M1431-5G
TCM1431-25GEA 150.1 CAD
TCM1431-25G TCM1431-5G
2'-Methylformanilide ≥98.0%
2'-Methylformanilide

CAS Number: 94-69-9
MDL Number: MFCD00014122
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 61


Other Product Names / Synonyms: N-Formyl-2-methylaniline

Formula: C₈H₉NO
MW: 135.17
Melting Pt: 61 °C
MDL Number: MFCD00014122
CAS Number: 94-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26b168bb46641bf599b8d4e1a238c3edc2c9ce8&vid=c26b168bb46641bf599b8d4e1a238c3edc2c9ce8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR