6-Methylindole ≥98.0%

Supplier: TCI America

M1430-5G M1430-1G
TCM1430-5GEA 101.12 CAD
TCM1430-5G TCM1430-1G
6-Methylindole ≥98.0%
6-Methylindole

CAS Number: 3420-02-8
MDL Number: MFCD00005682
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 29
Storage Temperature: 0-10°C

Formula: C₉H₉N
Flash Pt: 110 °C
MDL Number: MFCD00005682
CAS Number: 3420-02-8
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=714f7a434d524f0e4886f35de7dfa70645049f61&vid=714f7a434d524f0e4886f35de7dfa70645049f61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR