Methyl-3-methoxy-4-methylbenzoate ≥98.0%

Supplier: TCI America

M1427-25G M1427-5G
TCM1427-25GEA 368.14 CAD
TCM1427-25G TCM1427-5G
Methyl-3-methoxy-4-methylbenzoate ≥98.0%
Methyl 3-methoxy-4-methylbenzoate

CAS Number: 3556-83-0
MDL Number: MFCD00082710
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 3-Methoxy-4-methylbenzoic Acid Methyl Ester, 3-Methoxy-p-toluic Acid Methyl Ester, 4-Methyl-m-anisic Acid Methyl Ester, Methyl 3-Methoxy-p-toluate, Methyl 4-Methyl-m-anisate

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00082710
CAS Number: 3556-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66138b1b675ac6831d2d4821b7bf51e6efbd8f0f&vid=66138b1b675ac6831d2d4821b7bf51e6efbd8f0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR