4-Methyl-m-anisic acid ≥98.0%
Supplier: TCI America
Synonyms:
3-Methoxy-4-methylbenzoic acid
M1426-25G
M1426-5G
TCM1426-25GEA
118.5
CAD
TCM1426-25G
TCM1426-5G
4-Methyl-m-anisic acid ≥98.0%
4-Methyl-m-anisic acid
CAS Number: 7151-68-0
MDL Number: MFCD00002566
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 3-Methoxy-p-toluic Acid, 4-Methyl-m-anisic Acid
Formula:
C₉H₁₀O₃ MW: 166.18 |
MDL Number:
MFCD00002566 CAS Number: 7151-68-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...