7-Methoxyflavone ≥98.0%
Supplier: TCI America
Synonyms:
7-Methoxy-2-phenyl-4H-1-benzopyran-4-one, 7-Methoxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Methoxy-2-phenylchromen-4-one, 7-Methoxy-2-phenyl-4H-chromen-4-one, 7-Methoxy-2-phenyl-4-benzopyrone
M1423-1G
M1423-5G
TCM1423-1GEA
84.37
CAD
TCM1423-1G
TCM1423-5G
7-Methoxyflavone ≥98.0%
7-Methoxyflavone
CAS Number: 22395-22-8
MDL Number: MFCD00017462
Molecular Formula: C16H12O3
Molecular Weight: 252.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 111
Formula:
C₁₆H₁₂O₃ MW: 252.27 g/mol Melting Pt: 111 °C |
MDL Number:
MFCD00017462 CAS Number: 22395-22-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...