6-Methoxy-2-naphthoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M1418-25G
M1418-5G
TCM1418-25GEA
505.6
CAD
TCM1418-25G
TCM1418-5G
6-Methoxy-2-naphthoic acid ≥97.0% (by GC, titration analysis)
6-Methoxy-2-naphthoic acid
CAS Number: 2471-70-7
MDL Number: MFCD00092750
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Formula:
C₁₂H₁₀O₃ MW: 202.21 g/mol |
MDL Number:
MFCD00092750 CAS Number: 2471-70-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...