2-Methyl-3-nitroanisole ≥97.0%

Supplier: TCI America

M1416-5G M1416-25G
TCM1416-5GEA 129.56 CAD
TCM1416-5G TCM1416-25G
2-Methyl-3-nitroanisole ≥97.0%
2-Methyl-3-nitroanisole

CAS Number: 4837-88-1
MDL Number: MFCD00007159
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 56
Flash Point (°C): 110


Other Product Names / Synonyms: 1-Methoxy-2-methyl-3-nitrobenzene, 2-Methyl-3-nitroanisole

Formula: C₈H₉NO₃
Melting Pt: 56 °C
Flash Pt: 110 °C
MDL Number: MFCD00007159
CAS Number: 4837-88-1
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=218c16d82e254a5b2481d01e3216e4072c3d5f9d&vid=04277ff3a1980470fdbf92ff1f2c9052cb2fb58a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR