6-Methoxyflavanone ≥98.0%
Supplier: TCI America
M1403-5G
M1403-1G
TCM1403-5GEA
200.66
CAD
TCM1403-5G
TCM1403-1G
6-Methoxyflavanone ≥98.0%
6-Methoxyflavanone
CAS Number: 3034-04-6
MDL Number: MFCD00017484
Molecular Formula: C16H14O3
Molecular Weight: 254.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 142
Formula:
C₁₆H₁₄O₃ Melting Pt: 142 °C |
MDL Number:
MFCD00017484 CAS Number: 3034-04-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...