2-Methoxy-3-nitropyridine ≥98.0%

Supplier: TCI America

M1395-5G M1395-25G
TCM1395-5GEA 171.9 CAD
TCM1395-5G TCM1395-25G
2-Methoxy-3-nitropyridine ≥98.0%
2-Methoxy-3-nitropyridine

CAS Number: 20265-35-4
MDL Number: MFCD00023459
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56

Formula: C₆H₆N₂O₃
Melting Pt: 56 °C
MDL Number: MFCD00023459
CAS Number: 20265-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a19a3367e253e19f31cadb2bff6c06f5b1b550a2&vid=a19a3367e253e19f31cadb2bff6c06f5b1b550a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR