3-Methylbenzofuran-2-carboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1392-1G M1392-5G
TCM1392-1GEA 63.2 CAD
TCM1392-1G TCM1392-5G
3-Methylbenzofuran-2-carboxylic acid ≥98.0% (by titrimetric analysis)
3-Methylbenzofuran-2-carboxylic acid

CAS Number: 24673-56-1
MDL Number: MFCD00060511
Molecular Formula: C10H8O3
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 193

Formula: C₁₀H₈O₃
Melting Pt: 193 °C
MDL Number: MFCD00060511
CAS Number: 24673-56-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77476d507aa85b394c9023155286893721ef0c20&vid=77476d507aa85b394c9023155286893721ef0c20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR