4-(4-Methylphenyl)-4-oxobutyric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Oxo-4-(p-tolyl)butanoic acid, 3-(4-Methylbenzoyl)propionic Acid, 4-(p-Tolyl)-4-oxobutyric Acid

M1377-25G M1377-5G
TCM1377-25GEA 331.8 CAD
TCM1377-25G TCM1377-5G
4-(4-Methylphenyl)-4-oxobutyric acid ≥98.0% (by GC, titration analysis)
4-(4-Methylphenyl)-4-oxobutyric acid

CAS Number: 4619-20-9
MDL Number: MFCD00020541
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 128


Other Product Names / Synonyms: 3-(4-Methylbenzoyl)propionic Acid, 4-(p-Tolyl)-4-oxobutyric Acid

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00020541
CAS Number: 4619-20-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=496387cb5f96c5ab557c8f4c3c051f2a3c0f0348&vid=496387cb5f96c5ab557c8f4c3c051f2a3c0f0348&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR