Methyl-3-methoxy-2-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: 3-Methoxy-2-nitrobenzoic acid methyl ester, Methyl 3-methoxy-2-nitrobenzoate

M1364-1G M1364-5G
TCM1364-1GEA 71.1 CAD
TCM1364-1G TCM1364-5G
Methyl-3-methoxy-2-nitrobenzoate ≥98.0%
Methyl 3-methoxy-2-nitrobenzoate

CAS Number: 5307-17-5
MDL Number: MFCD00051968
Molecular Formula: C9H9NO5
Molecular Weight: 211.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 142


Other Product Names / Synonyms: 3-Methoxy-2-nitrobenzoic Acid Methyl Ester, Methyl 2-Nitro-m-anisate, 2-Nitro-m-anisic Acid Methyl Ester

Formula: C₉H₉NO₅
MW: 211.17 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00051968
CAS Number: 5307-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec0f90b6bdb02365113b3e8c9613a3e80eb50c72&vid=ec0f90b6bdb02365113b3e8c9613a3e80eb50c72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR