α-Methylcinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methyl-3-phenyl-2-propenoic acid

M1336-25G M1336-5G
TCM1336-25GEA 128.93 CAD
TCM1336-25G TCM1336-5G
α-Methylcinnamic acid ≥98.0% (by GC, titration analysis)
α-Methylcinnamic acid

CAS Number: 1199-77-5
MDL Number: MFCD00002652
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 288
Melting point (°C): 80

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 288 °C
Melting Pt: 80 °C
MDL Number: MFCD00002652
CAS Number: 1199-77-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1aa5b39f283f93254fb5373ced07c4d62f1f1c3&vid=c1aa5b39f283f93254fb5373ced07c4d62f1f1c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR