Methyl methanesulfonylacetate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-(methylsulphonyl)acetate
, Methanesulfonylacetic acid Methyl Ester
, Methyl 2-(methylsulfonyl)acetate
, Methanesulphonylacetic acid Methyl Ester
M1332-5G
TCM1332-005GEA
227.84
CAD
TCM1332-005G
Methyl methanesulfonylacetate ≥98.0%
Methyl methanesulfonylacetate
CAS Number: 62020-09-1
MDL Number: MFCD00051842
Molecular Formula: C4H8O4S
Molecular Weight: 152.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62
Other Product Names / Synonyms: Methanesulfonylacetic Acid Methyl Ester
Formula:
C₄H₈O₄S MW: 152.16 g/mol Melting Pt: 62 °C |
MDL Number:
MFCD00051842 CAS Number: 62020-09-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...