Methyl (3-methoxyphenyl)acetate ≥98.0%

Supplier: TCI America

M1331-25G M1331-5G
TCM1331-025GEA 186.44 CAD
TCM1331-025G TCM1331-5G
Methyl (3-methoxyphenyl)acetate ≥98.0%
Methyl (3-methoxyphenyl)acetate

CAS Number: 18927-05-4
MDL Number: MFCD00017205
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 3-Methoxyphenylacetic Acid Methyl Ester

Formula: C₁₀H₁₂O₃
Density: 1.12 (20 °C)
MDL Number: MFCD00017205
CAS Number: 18927-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef2b9f891630246c226ae57c9e90104ed0097687&vid=ef2b9f891630246c226ae57c9e90104ed0097687&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR