2-Mercapto-5-nitrobenzimidazole ≥98.0% (by HPLC)

Supplier: TCI America

M1325-5G M1325-25G
TCM1325-5GEA 224.36 CAD
TCM1325-5G TCM1325-25G
2-Mercapto-5-nitrobenzimidazole ≥98.0% (by HPLC)
2-Mercapto-5-nitrobenzimidazole

CAS Number: 6325-91-3
MDL Number: MFCD00804727
Molecular Formula: C7H5N3O2S
Molecular Weight: 195.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 5-Nitro-2-benzimidazolethiol

Formula: C₇H₅N₃O₂S
MDL Number: MFCD00804727
CAS Number: 6325-91-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b42ac5c79705245282520bcf5a7a024a0a06f0f1&vid=b42ac5c79705245282520bcf5a7a024a0a06f0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR