3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98.0% stabilized

Supplier: TCI America

Synonyms: 3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl methacrylate, 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane

M1324-25ML M1324-100ML
TCM1324-025MLEA 74.26 CAD
TCM1324-025ML TCM1324-100ML
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98.0% stabilized
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate

(stabilized with MEHQ)
CAS Number: 17096-07-0
MDL Number: MFCD00053871
Molecular Formula: C16H38O5Si4
Molecular Weight: 422.82
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 200
Specific Gravity (20/20): 0.93


Product Categories: Materials Chemistry, Silane Coupling Agents, Monomers
Other Product Names / Synonyms: 3-(Methacryloyloxy)propyltris(trimethylsilyloxy)silane, Methacrylic Acid 3-[Tris(trimethylsilyloxy)silyl]propyl Ester

Formula: C₁₆H₃₈O₅Si₄
MW: 422.82 g/mol
Boiling Pt: 115 °C
Density: 0.93 (20 °C)
Flash Pt: 200 °C
MDL Number: MFCD00053871
CAS Number: 17096-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d554aa93be22da87365d95a5221108f2bf9c09&vid=66d554aa93be22da87365d95a5221108f2bf9c09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR