2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol] ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2'-Methylenebis[4-methyl-6-(1-methylcyclohexyl)phenol], Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane, 2,2'-Dihydroxy-3,3'-bis(alpha-methylcyclohexyl)-5,5'-dimethyldiphenylmethane

M1301-25G M1301-500G
TCM1301-025GEA 45.82 CAD
TCM1301-025G TCM1301-500G
2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol] ≥95.0% (by titrimetric analysis)
2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol]

CAS Number: 77-62-3
MDL Number: MFCD00151797
Molecular Formula: C29H40O2
Molecular Weight: 420.64
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 135
Flash Point (°C): 222


Other Product Names / Synonyms: Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane, 2,2'-Dihydroxy-3,3'-bis(alpha-methylcyclohexyl)-5,5'-dimethyldiphenylmethane, 2,2'-Methylenebis[4-methyl-6-(1-methylcyclohexyl)phenol]

Formula: C₂₉H₄₀O₂
MW: 420.64 g/mol
Melting Pt: 135 °C
Flash Pt: 222 °C
MDL Number: MFCD00151797
CAS Number: 77-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a9dc06e66cf5b24950276931d6ae5ca3ff162e7&vid=4a9dc06e66cf5b24950276931d6ae5ca3ff162e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR